missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLIC1 Protein

A cDNA sequence encoding the CLIC1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11447-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917298

  • 160.00€ / 25 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

CLIC1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CLIC1 Human solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
GSSHHHHHH SSGLVPRGSH MAEEQPQVEL FVKAGSDGAK IGNCPFSQRL FMVLWLKGVT FNVTTVDTKR RTETVQKLCP GGQLPFLLYG TEVHTDTNKI EEFLEAVLCP PRYPKLAALN PESNTAGLDI FAKFSAYIKN SNPALNDNLE KGLLKALKVL DNYLTSPLPE EVDETSAEDE GVSQRKFLDG NELTLADCNL LPKLHIVQVV CKKYRGFTIP EAFRGVHRYL SNAYAREEFA STCPDDEEIE LAYEQVAKAL K
Videos