missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clavesin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Clavesin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7684-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001010852.2 | |
Clavesin-2 Protein | |
Research Use Only | |
CLVS2 | |
Recombinant Protein | |
E. coli | |
MTHLQAGLSP ETLEKARLEL NENPDTLHQD IQEVRDMVIT RPDIGFLRTD DAFILRFLRA RKFHHFEAFR LLAQYFEYRQ QNLDMFKSFK ATDPGIKQAL KDGFPGGLAN LDHYGRKILV LFAANWDQSR YTLVDILRAI LLSLEAMIED PELQVNGFVL IIDWSNFTFK QASKLTPSML RLAIEGLQDS FPARFGGIHF VNQPWYIHAL YTVIRPFLKE KTRKRIFLHG NNLNSLHQLI HPEILPSEFG GMLPPYDMGT WARTLLDHEY DDDSEYNVDS YSMPVKEVEK ELSPKSMKRS QSVVDPTVLK RMDKNEEENM QPLLSLD | |
Greater than 90% as determined by SDS-PAGE. |
134829 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |