missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKMT1 Protein

A cDNA sequence encoding the CKMT1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10555-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914238

  • 160.00€ / 50 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

1158
Greater than 95.0% as determined bya) Analysis by RP-HPLCb) Analysis by SDS-PAGE
Research Use Only
CKM
Recombinant Protein
Pichia Pastoris
MPFGNTHNKF KLNYKPEEEY PDLSKHNNHM AKVLTLELYK KLRDKETPSG FTVDDVIQTG VDNPGHPFIM TVGCVAGDEE SYEVFKELFD PIISDRHGGY KPTDKHKTDL NHENLKGGDD LDPNYVLSSR VRTGRSIKGY TLPPHCSRGE RRAVEKLSVE ALNSLTGEFK GKYYPLKSMT EKEQQQLIDD HFLFDKPVSP LLLASGMARD WPDARGIWHN DNKSFLVWVN EEDHLRVISM EKGGNMKEVF RRFCVGLQKI EEIFKKAGHP FMWNQHLGYV LTCPSNLGTG LRGGVHVKLA HLSKHPKFEE ILTRLRLQKR GTGGVDTAAV GSVFDVSNAD RLGSSEVEQV QLVVDGVKLM VEMEKKLEKG QSIDDMIPAQ K
CKMT1 Protein
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
The biological activity measured by the enzymatic activity of Creatine phosphokinase procedure No. 45-UV, 1IU-1 micromole creatine phosphate was 537IU/mg at 37 degrees celsius corresponding to a Specific Activity of 1,863ng/ml.
Human
Untagged
CKMT1 contains 0.01M Tris-HCl, 0.075M NaCl, 10mM beta-mercaptoethanol, 50% glycerol, 0.1% sodium azide, pH 7.2.
Videos