missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKB Protein

A cDNA sequence encoding the CKB was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10552-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914228

  • 1083.00€ / 1 miligramo


Fecha estimada de envío
16-07-2019
Toevoegen aan winkelmandje

Specificatie

Specificatie

1152
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
CKB
Recombinant Protein
Pichia Pastoris
MPFSNSHNAL KLRFPAEDEF PDLSAHNNHM AKVLTPELYA ELRAKSTPSG FTLDDVIQTG VDNPGHPYIM TVGCVAGDEE SYEVFKDLFD PIIEDRHGGY KPSDEHKTDL NPDNLQGGDD LDPNYVLSSR VRTGRSIRGF CLPPHCSRGE RRAIEKLAVE ALSSLDGDLA GRYYALKSMT EAEQQQLIDD HFLFDKPVSP LLLASGMARD WPDARGIWHN DNKTFLVWVN EEDHLRVISM QKGGNMKEVF TRFCTGLTQI ETLFKSKDYE FMWNPHLGYI LTCPSNLGTG LRAGVHIKLP NLGKHEKFSE VLKRLRLQKR GTGGVDTAAV GGVFDVSNAD RLGFSEVELV QMVVDGVKLL IEMEQRLEQG QAIDDLMPAQ K
CKB Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The biological activity measured by the enzymatic activity of Creatine phosphokinase procedure No. 45-UV, 1IU-1 micromole creatine phosphate was 854 IU/mg at 37 degrees celsius corresponding to a Specific Activity of 1,171ng/ml.
Human
Untagged
CKBB Human contains 10Mm Bis-Tris-HCl pH 6.0, 50% glycerol, 0.5mM EDTA and 0.5mM DTT.
Video's