missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CIB2 Protein

A cDNA sequence encoding the CIB2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11429-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917238

  • 3091.00€ / 1 miligramo


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

10518
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
CIB2
Human
His
The CIB2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT, 100mM NaCl and 1mM PMSF.
CIB2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGNKQTIFTE EQLDNYQDCT FFNKKDILKL HSRFYELAPN LVPMDYRKSP IVHVPMSLII QMPELRENPF KERIVAAFSE DGEGNLTFND FVDMFSVLCE SAPRELKANY AFKIYDFNTD NFICKEDLEL TLARLTKSEL DEEEVVLVCD KVIEEADLDG DGKLGFADFE DMIAKAPDFL STFHIRI
Videos
Documentos