missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Chymotrypsin-like elastase family member 2A Protein

A DNA sequence encoding the Homo sapiens (Human) Chymotrypsin-like elastase family member 2A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5816-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917969

  • 218.00€ / 10 microgramos


Fecha estimada de envío
18-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_254275.1
Chymotrypsin-like elastase family member 2A Protein
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
63036
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Cela2a
Recombinant Protein
Yeast
VVGGEEARPN SWPWQVSLQY SSNGKWYHTC GGSLIANSWV LTAAHCISSS RTYRVGLGRH NLYVAESGSL AVSVSKIVVH KDWNSNQISK GNDIALLKLA NPVSLTDKIQ LACLPPAGTI LPNNYPCYVT GWGRLQTNGA VPDVLQQGRL LVVDYATCSS SAWWGSSVKT SMICAGGDGV ISSCNGDSGG PLNCQASDGR WQVHGIVSFG SRLGCNYYHK PSVFTRVSNY IDWINSVIAN N
Videos