missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Chitinase domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Chitinase domain-containing protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8193-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053191

  • 287.00€ / 10 microgramos


Fecha estimada de envío
24-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001136146.1
Chitinase domain-containing protein 1
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
66005
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Chid1
Recombinant Protein
Yeast
TLSKSDAKKA ASKTLLEKSQ FSDKPVQDRG LVVTDLKAES VVLEHRSYCS AKARDRHFAG DVLGYVTPWN SHGYDVTKVF GSKFTQISPV WLQLKRRGRE MFEVTGLHDV DQGWMRAVRK HAKGLHIVPR LLFEDWTYDD FRNVLDSEDE IEELSKTVVQ VAKNQHFDGF VVEVWNQLLS QKRVGLIHML THLAEALHQA RLLALLVIPP AITPGTDQLG MFTHKEFEQL APVLDGFSLM TYDYSTAHQP GPNAPLSWVR ACVQVLDPKS KWRSKILLGL NFYGMDYATS KDAREPVVGA RYIQTLKDHR PRMVWDSQAS EHFFEYKKSR SGRHVVFYPT LKSLQVRLEL ARELGVGVSI WELGQGLDYF YDLL
Videos
Documentos