missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHCHD7 Protein
A cDNA sequence encoding the CHCHD7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11403-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
79145 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMHQTRTG KKTVRMPSVT QRLRDPDINP CLSESDASTR CLDENNYDRE RCSTYFLRYK NCRRFWNSIV MQRRKNGVKP FMPTAAERDE ILRAVGNMPY | |
Greater than 90% as determined by SDS-PAGE. |
CHCHD7 Protein | |
Research Use Only | |
CHCHD7 | |
Human | |
His | |
CHCHD7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |