missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cell division cycle protein 23 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Cell division cycle protein 23 homolog, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8237-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053471

  • 234.00€ / 50 microgramos


Fecha estimada de envío
16-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_004652.2
Cell division cycle protein 23 homolog Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
8697
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CDC23
Recombinant Protein
E. coli
AAVAPVLSIN SDFSDLREIK KQLLLIAGLT RERGLLHSSK WSAELAFSLP ALPLAELQPP PPITEEDAQD MDAYTLAKAY FDVKEYDRAA HFLHGCNSKK AYFLYMYSRY LSGEKKKDDE TVDSLGPLEK GQVKNEALRE LRVELSKKHQ ARELDGFGLY LYGVVLRKLD LVKEAIDVFV EATHVLPLHW GAWLELCNLI TDKEMLKFLS LPDTWMKEFF LAHIYTELQL IEEALQKYQN LIDVGFSKSS YIVSQIAVAY HNIRDIDKAL SIFNELRKQD PYRIENMDTF SNLLYVRSMK SELSYLAHNL CEIDKYRVET CCVIGNYYSL RSQHEKAALY FQRALKLNPR YLGAWTLMGH EYMEMKNTSA AIQAYRHAIE VNKRDYRAWY GLGQTYEILK MPFYCLYYYR RAHQLRPNDS RMLVALGECY EKLNQLVEAK KCYWRAYAVG DVEKMALVKL AKLHEQLTES EQAAQCYIKY IQDIYSCGEI VEHLEESTAF RYLAQYYFKC KLWDEASTCA QKCCAFNDTR EEGKALLRQI LQLRNQGETP TTEVPAPFFL PASLSANNTP TRRVSPLN
Videos
Documentos