missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDKN2AIP N-terminal-like Protein

A DNA sequence encoding the Homo sapiens (Human) CDKN2AIP N-terminal-like protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5808-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15907909

  • 437.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_542387.1
CDKN2AIP N-terminal-like Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
91368
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CDKN2AIPNL
Recombinant Protein
E. coli
MVGGEAAAAV EELVSGVRQA ADFAEQFRSY SESEKQWKAR MEFILRHLPD YRDPPDGSGR LDQLLSLSMV WANHLFLGCS YNKDLLDKVM EMADGIEVED LPQFTTRSEL MKKHQS
Videos