missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK5 Protein
A DNA sequence encoding the Homo sapiens (Human) CDK5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5806-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001157882.1 | |
CDK5 Protein | |
Research Use Only | |
CDK5 | |
Recombinant Protein | |
E. coli | |
MQKYEKLEKI GEGTYGTVFK AKNRETHEIV ALKRVRLDDD DEGVPSSALR EICLLKELKH KNIVRLHDVL HSDKKLTLVF EFCDQDLKKY FDSCNGDLDP EIVKSFLFQL LKGLGFCHSR NVLHRDLKPQ NLLINRNGEL KLADFGLARA FGIPVRCYSA EVVTLWYRPP DVLFGAKLYS TSIDMWSAGC IFAELANAGR PLFPGNDVDD QLKRIFRLLG TPTEEQWPSM TKLPDYKPYP MYPATTSLVN VVPKLNATGR DLLQNLLKCN PVQRISAEEA LQHPYFSDFC PP | |
Greater than 90% as determined by SDS-PAGE. |
1020 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |