missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK2 / p33 Protein
A DNA sequence encoding the Homo sapiens (Human) CDK2 / p33, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5804-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001277159.1 | |
CDK2 / p33 Protein | |
Research Use Only | |
CDK2 | |
Recombinant Protein | |
E. coli | |
MENFQKVEKI GEGTYGVVYK ARNKLTGEVV ALKKIRLDTE TEGVPSTAIR EISLLKELNH PNIVKLLDVI HTENKLYLVF EFLHQDLKKF MDASALTGIP LPLIKSYLFQ LLQGLAFCHS HRVLHRDLKP QNLLINTEGA IKLADFGLAR AFGVPVRTYT HEVVTLWYRA PEILLGCKYY STAVDIWSLG CIFAEMVTRR ALFPGDSEID QLFRIFRTLG TPDEVVWPGV TSMPDYKPSF PKWARQDFSK VVPPLDEDGR SLLSQMLHYD PNKRISAKAA LAHPFFQDVT KPVPHLRL | |
Greater than 90% as determined by SDS-PAGE. |
1017 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |