missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD81 / TAPA-1 Protein

A DNA sequence encoding the Homo sapiens (Human) CD81 / TAPA-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5797-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15907839

  • 1926.00€ / 1 miligramo


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001284578.1
CD81 / TAPA-1 Protein
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
975
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cd81
Recombinant Protein
E. coli
FVNKDQIAKD VKQFYDQALQ QAVVDDDANN AKAVVKTFHE TLDCCGSSTL TALTTSVLKN NLCPSGSNII SNLFKEDCHQ KIDDLFSGK
Videos
Documentos