missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD7 Protein

A DNA sequence encoding the Homo sapiens (Human) CD7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5794-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917819

  • 218.00€ / 10 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_006128.1
CD7 Protein
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
924
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD7
Recombinant Protein
Yeast
AQEVQQSPHC TTVPVGASVN ITCSTSGGLR GIYLRQLGPQ PQDIIYYEDG VVPTTDRRFR GRIDFSGSQD NLTITMHRLQ LSDTGTYTCQ AITEVNVYGS GTLVLVTEEQ SQGWHRCSDA PPRASALPAP PTGSALPDPQ TASALPDPPA ASALP
Videos