missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD68 Protein
A cDNA sequence encoding the CD68 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11343-HIS-EC-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
968 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRLAVLFSGA LLGLLAAQGT GNDCPHKKSA TLLPSFTVTP TVTESTGTTS HRTTKSHKTT THRTTTTGTT SHGPTTATHN PTTTSHGNVT VHPTSNSTAT SQGPSTATHS PATTSHGNAT VHPTSNSTAT SPGFTSSAHP EPPPPSPSPS PTSKETIGDY TWTNGSQPCV HLQAQIQIRV MYTTQGGGEA WGISVLNPNK TKVQGSCEGA HPHLLLSFPY GHLSFGFMQD LQQKVVYLSY MAVEYNVSFP HAAQWTFSAQ NASLRDLQAP LGQSFSCSNS SIILSPAVHL DLLSLRLQAA QLPHTGVFGQ SFSCPSDRSI LLPLIIGLIL LGLLALVLIA FCIIRRRPSA YQAL |
CD68 Protein | |
Research Use Only | |
CD68 | |
Human | |
His | |
CD68 at 100microg/1ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA, 20% Glycerol. |