missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD40L / CD154 / TNFSF5 Protein
A DNA sequence encoding the Human CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10359-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000065.1 | |
CD40L / CD154 / TNFSF5 Protein | |
Research Use Only | |
CD40LG | |
The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 5-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.0, lyophilized |
959 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL | |
Greater than 95% as determined by SDS-PAGE and HPLC. |