missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD164L2 Protein
A cDNA sequence encoding the CD164L2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11306-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
388611 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGKGARGF GRGALIRLNI WPAVQGACKQ LEVCEHCVEG DRARNLSSCM WEQCRPEEPG HCVAQSEVVK EGCSIYNRSE ACPAAHHHPT YEPKTVTTGS PPVPEAHSPG FDGAS | |
Greater than 90% as determined by SDS-PAGE. |
CD164L2 Protein | |
Research Use Only | |
CD164L2 | |
Human | |
His | |
CD164L2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |