missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD146 / MCAM Protein

A DNA sequence encoding the Homo sapiens (Human) CD146 / MCAM, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8689-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055971

  • 234.00€ / 50 microgramos


Fecha estimada de envío
28-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_006491.2
CD146 / MCAM Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
4162
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MCAM
Recombinant Protein
E. coli
LSQSQGNLSH VDWFSVHKEK RTLIFRVRQG QGQSEPGEYE QRLSLQDRGA TLALTQVTPQ DERIFLCQGK RPRSQEYRIQ LRVYKAPEEP NIQVNPLGIP VNSKEPEEVA TCVGRNGYPI PQVIWYKNGR PLKEEKNRVH IQSSQTVESS GLYTLQSILK AQLVKEDKDA QFYCELNYRL PSGNHMKESR EVTVPVFYPT EKVWLEVEPV GMLKEGDRVE IRCLADGNPP PHFSISKQNP STREAEEETT NDNGVLVLEP ARKEHSGRYE CQGLDLDTMI SLLSEPQELL VNYVSDVRVS PAAPERQEGS SLTLTCEAES SQDLEFQWLR EETGQVLERG PVLQLHDLKR EAGGGYRCVA SVPSIPGLNR TQLVNVAIFG PPWMAFKERK VWVKENMVLN LSCEASGHPR PTISWNVNGT ASEQDQDPQR VLSTLNVLVT PELLETGVEC TASNDLGKNT SILFLELVNL TTLTPDSNTT TGLSTSTASP HTRANSTSTE RKLPEPESRG VVIVAVIVCI LVLAVLGAVL YFLYKKGKLP CRRSGKQEIT LPPSRKSELV VEVKSDKLPE EMGLLQGSSG DKRAPGDQGE KYIDLRH
Videos
Documentos