missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD137 / 4-1BB / TNFRSF9 Protein
A DNA sequence encoding the Human CD137 / 4-1BB / TNFRSF9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10333-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001552.2 | |
CD137 / 4-1BB / TNFRSF9 Protein | |
Research Use Only | |
TNFRSF9 | |
The biological activity is determined by its inhibitory effect of IL-8 production using human peripheral blood mononuclear cells. About 90 % of inibition was seen using a concentration of 1 ug for both 4-1BB Ligand and 4-1BB Receptor. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3604 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ERTRSLQDPC SNCPAGTFCD NNRNQICSPC PPNSFSSAGG QRTCDICRQC KGVFRTRKEC SSTSNAECDC TPGFHCLGAG CSMCEQDCKQ GQELTKKGCK DCCFGTFNDQ KRGICRPWTN CSLDGKSVLV NGTKERDVVC GPSPADLSPG ASSVTPPAPA REPGHS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |