missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD112 / Nectin-2 / PVRL2 Protein

A DNA sequence encoding the Homo sapiens (Human) CD112 / Nectin-2 / PVRL2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7863-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051251

  • 234.00€ / 50 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001036189.1
CD112 / Nectin-2 / PVRL2 Protein
50 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
5819
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PVRL2
Recombinant Protein
E. coli
QDVRVQVLPE VRGQLGGTVE LPCHLLPPVP GLYISLVTWQ RPDAPANHQN VAAFHPKMGP SFPSPKPGSE RLSFVSAKQS TGQDTEAELQ DATLALHGLT VEDEGNYTCE FATFPKGSVR GMTWLRVIAK PKNQAEAQKV TFSQDPTTVA LCISKEGRPP ARISWLSSLD WEAKETQVSG TLAGTVTVTS RFTLVPSGRA DGVTVTCKVE HESFEEPALI PVTLSVRYPP EVSISGYDDN WYLGRTDATL SCDVRSNPEP TGYDWSTTSG TFPTSAVAQG SQLVIHAVDS LFNTTFVCTV TNAVGMGRAE QVIFVRETPN TAGAGATGG
Videos
Documentos