missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCN3 / NOV Protein
A DNA sequence encoding the Human CCN3 / NOV, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10388-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002505.1 | |
CCN3 / NOV Protein | |
Research Use Only | |
NOV | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1.0 ug/ml, corresponding to a specific activity of > 1000 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.0, lyophilized |
4856 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MQVAATQRCP PQCPGRCPAT PPTCAPGVRA VLDGCSCCLV CARQRGESCS DLEPCDESSG LYCDRSADPS NQTGICTAVE GDNCVFDGVI YRSGEKFQPS CKFQCTCRDG QIGCVPRCQL DVLLPEPNCP APRKVEVPGE CCEKWICGPD EEDSLGGLTL AAYRPEATLG VEVSDSSVNC IEQTTEWTAC SKSCGMGFST RVTNRNRQCE MLKQTRLCMV RPCEQEPEQP TDKKGKKCLR TKKSLKAIHL QFKNCTSLHT YKPRFCGVCS DGRCCTPHNT KTIQAEFQCS PGQIVKKPVM VIGTCTCHTN CPKNNEAFLQ ELELKTTRGK M | |
Greater than 95% as determined by SDS-PAGE and HPLC. |