missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCN3 / NOV Protein

A DNA sequence encoding the Human CCN3 / NOV, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10388-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913708

  • 1510.00€ / 250 microgramos


Fecha estimada de envío
22-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002505.1
CCN3 / NOV Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MQVAATQRCP PQCPGRCPAT PPTCAPGVRA VLDGCSCCLV CARQRGESCS DLEPCDESSG LYCDRSADPS NQTGICTAVE GDNCVFDGVI YRSGEKFQPS CKFQCTCRDG QIGCVPRCQL DVLLPEPNCP APRKVEVPGE CCEKWICGPD EEDSLGGLTL AAYRPEATLG VEVSDSSVNC IEQTTEWTAC SKSCGMGFST RVTNRNRQCE MLKQTRLCMV RPCEQEPEQP TDKKGKKCLR TKKSLKAIHL QFKNCTSLHT YKPRFCGVCS DGRCCTPHNT KTIQAEFQCS PGQIVKKPVM VIGTCTCHTN CPKNNEAFLQ ELELKTTRGK M
4856
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
NOV
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1.0 ug/ml, corresponding to a specific activity of > 1000 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.0, lyophilized
Videos
Documentos