missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL8 / MCP-2 Protein

A DNA sequence encoding the Human CCL8 / MCP-2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10231-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913168

  • 299.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_005614.2
CCL8 / MCP-2 Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
6355
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL8
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-100 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos