missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL5 / RANTES Protein

A DNA sequence encoding the Human CCL5 / RANTES, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10229-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923158

Cantidad Precio
1 1095.00€ / 100 microgramos
4 1020.98€ / 100 microgramos
Fecha estimada de envío
14-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_002976.2
CCL5 / RANTES Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
6352
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL5
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 1.0-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos