missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL27 / CTACK Protein
A DNA sequence encoding the Human CCL27 / CTACK, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10242-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006655.1 | |
CCL27 / CTACK Protein | |
Research Use Only | |
CCL27 | |
The biological activity determined by a chemotaxis bioassay using CCR10 transfected BaF3 cells is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
10850 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
FLLPPSTACC TQLYRKPLSD KLLRKVIQVE LQEADGDCHL QAFVLHLAQR SICIHPQNPS LSQWFEHQER KLHGTLPKLN FGMLRKMG | |
Greater than 96% as determined by SDS-PAGE and HPLC. |