missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL27 / CTACK Protein

A DNA sequence encoding the Human CCL27 / CTACK, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10242-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923208

  • 1153.00€ / 250 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_006655.1
CCL27 / CTACK Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
FLLPPSTACC TQLYRKPLSD KLLRKVIQVE LQEADGDCHL QAFVLHLAQR SICIHPQNPS LSQWFEHQER KLHGTLPKLN FGMLRKMG
10850
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL27
The biological activity determined by a chemotaxis bioassay using CCR10 transfected BaF3 cells is in a concentration range of 10-100 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos