missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL26 / Eotaxin 3 Protein
Recombinant Protein
Marca: enQuireBio™ QP5304-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006063.1 | |
CCL26 / Eotaxin 3 Protein | |
Research Use Only | |
CCL26 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
10344 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
TRGSDISKTC CFQYSHKPLP WTWVRSYEFT SNSCSQRAVI FTTKRGKKVC THPRKKWVQK YISLLKTPKQ L | |
Greater than 95% as determined by SDS-PAGE. |