missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL22 / MDC Protein
A DNA sequence encoding the Human CCL22 / MDC, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10237-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002981.2 | |
CCL22 / MDC Protein | |
Research Use Only | |
CCL22 | |
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6367 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPYGANMEDS VCCRDYVRYR LPLRVVKHFY WTSDSCPRPG VVLLTFRDKE ICADPRVPWV KMILNKLSQ | |
Greater than 97% as determined by SDS-PAGE and HPLC. |