missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL20 / MIP-3 alpha Protein

A DNA sequence encoding the Human CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10235-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923178

  • 668.00€ / 100 microgramos


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001123518.1
CCL20 / MIP-3 alpha Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKNM
6364
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL20
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-50 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos