missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL15 / MIP-5 / MIP-1 delta Protein

A DNA sequence encoding the Human CCL15 / MIP-5 / MIP-1 delta, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10200-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933078

  • 105.00€ / 5 microgramos


Fecha estimada de envío
06-08-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_116741.2
CCL15 / MIP-5 / MIP-1 delta Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SFHFAADCCT SYISQSIPCS LMKSYFETSS ECSKPGVIFL TKKGRQVCAK PSGPGVQDCM KKLKPYSI
6359
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL15
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 1.0-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos