missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL11 / Eotaxin Protein

A DNA sequence encoding the Homo sapiens (Human) CCL11 / Eotaxin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5770-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937649

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
29-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002977.1
CCL11 / Eotaxin Protein
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6356
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CCL11
Recombinant Protein
E. coli
GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
Videos
Documentos