missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC43 Protein
A cDNA sequence encoding the CCDC43 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11290-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CCDC43 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAPSEV AAIAPGEGDG GGGGFGSWLD GRLEALGVDR AVYGAYILGI LQEEEEEEKL DALQGILSAF LEEDSLLNIC KEIVERWSET QNVVTKVKKE DEVQAIATLI EKQAQIVVKP RMVSEEEKQR KAALLAQYAD VTDEEDEADE KDDSGATTMN IGSDKLLFRN TNVEDVLNAR KLERDSLRDE SQRKKEQDKL QRERDKLAKQ ERKEKEKKRT QRGERKR | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CCDC43 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |