missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CBX1 Protein
A cDNA sequence encoding the CBX1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11283-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CBX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGKKQNKKKV EEVLEEEEEE YVVEKVLDRR VVKGKVEYLL KWKGFSDEDN TWEPEENLDC PDLIAEFLQS QKTAHETDKS EGGKRKADSD SEDKGEESKP KKKKEESEKP RGFARGLEPE RIIGATDSSG ELMFLMKWKN SDEADLVPAK EANVKCPQVV ISFYEERLTW HSYPSEDDDK KDDKN | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CBX1 protein 1 mg/ml is supplied in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 2mM DTT and 20% Glycerol. |