missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Caspase-5 Protein

A DNA sequence encoding the Homo sapiens (Human) Caspase-5, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5763-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917579

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001129581.1
Caspase-5 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
838
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CASP5
Recombinant Protein
E. coli
DQKITSVKPL LQIEAGPPES AESTNILKLC PREEFLRLCK KNHDEIYPIK KREDRRRLAL IICNTKFDHL PARNGAHYDI VGMKRLLQGL GYTVVDEKNL TARDMESVLR AFAARPEHKS SDSTFLVLMS HGILEGICGT AHKKKKPDVL LYDTIFQIFN NRNCLSLKDK PKVIIVQACR GEKHGELWVR D
Videos