missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CASP3 / Caspase-3 / CASP-3 Protein

A DNA sequence encoding the Homo sapiens (Human) CASP3 / Caspase-3 / CASP-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8509-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055021

  • 183.00€ / 10 microgramos


Fecha estimada de envío
17-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_004337.2
CASP3 / Caspase-3 / CASP-3 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
836
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CASP3
Recombinant Protein
E. coli
SGISLDNSYK MDYPEMGLCI IINNKNFHKS TGMTSRSGTD VDAANLRETF RNLKYEVRNK NDLTREEIVE LMRDVSKEDH SKRSSFVCVL LSHGEEGIIF GTNGPVDLKK ITNFFRGDRC RSLTGKPKLF IIQACRGTEL DCGIETD
Videos
Documentos