missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein

A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5880-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918419

  • 1453.00€ / 500 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001276841.1
Casein kinase I isoform epsilon Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
102800317
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSNK1E
Recombinant Protein
Yeast
MELRVGNKYR LGRKIGSGSF GDIYLGANIA SGEEVAIKLE CVKTKHPQLH IESKFYKMMQ GGVGIPSIKW CGAEGDYNVM VMELLGPSLE DLFNFCSRKF SLKTVLLLAD QMISRIEYIH SKNFIHRDVK PDNFLMGLGK KGNLVYIIDF GLAKKYRDAR THQHIPYREN KNLTGTARYA SINTHLGIEQ SRRDDLESLG YVLMYFNLGS LPWQGLKAAT KRQKYERISE KKMSTPIEVL CKGYPSEFST YLNFCRSLRF DDKPDYSYLR QLFRNLFHRQ GFSYDYVFDW NMLKFGAARN PEDVDRERRE HEREERMGQL RGSATRALPP GPPTGATANR LRSAAEPVAS TPASRIQPAG NTSPRAISRV DRERKVSMRL HRGAPANVSS SDLTGRQEVS RIPASQTSVP FDHLGK
Videos