missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein

A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8358-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054211

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001276841.1
Casein kinase I isoform epsilon Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
102800317
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSNK1E
Recombinant Protein
E. coli
SGSFGDIYLG ANIASGEEVA IKLECVKTKH PQLHIESKFY KMMQGGVGIP SIKWCGAEGD YNVMVMELLG PSLEDLFNFC SRKFSLKTVL LLADQMISRI EYIHSKNFIH RDVKPDNFLM GLGKKGNLVY IIDFGLAKKY RDARTHQHIP YRENKNLTGT ARYASINTHL GIEQSRRDDL ESLGYVLMYF NLGSLPWQGL KAATKRQKYE RISEK
Videos
Documentos