missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Carboxypeptidase A6 Protein

A DNA sequence encoding the Homo sapiens (Human) Carboxypeptidase A6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5857-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918279

  • 253.00€ / 50 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_065094.3
Carboxypeptidase A6 Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
57094
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CPA6
Recombinant Protein
E. coli
SLSGYNYEVY HSLEEIQNWM HHLNKTHSGL IHMFSIGRSY EGRSLFILKL GRRSRLKRAV WIDCGIHARE WIGPAFCQWF VKEALLTYKS DPAMRKMLNH LYFYIMPVFN VDGYHFSWTN DRFWRKTRSR NSRFRCRGVD ANRNWKVKWC DEGASMHPCD DTYCGPFPES EPEVKAVANF LRKHRKHIRA YLSFHAYAQM LLYPYSYKYA TIPNFRCVES AAYKAVNALQ SVYGVRYRYG PASTTLYVSS GSSMDWAYKN GIPYAFAFEL RDTGYFGFLL PEMLIKPTCT ETMLAVKNIT MHLLKKCP
Videos