missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonyl reductase family member 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonyl reductase family member 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7969-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_116172.2 | |
Carbonyl reductase family member 4 Protein | |
Research Use Only | |
CBR4 | |
Recombinant Protein | |
E. coli | |
MDKVCAVFGG SRGIGRAVAQ LMARKGYRLA VIARNLEGAK AAAGDLGGDH LAFSCDVAKE HDVQNTFEEL EKHLGRVNFL VNAAGINRDG LLVRTKTEDM VSQLHTNLLG SMLTCKAAMR TMIQQQGGSI VNVGSIVGLK GNSGQSVYSA SKGGLVGFSR ALAKEVARKK IRVNVVAPGF VHTDMTKDLK EEHLKKNIPL GRFGETIEVA HAVVFLLESP YITGHVLVVD GGLQLIL | |
Greater than 80% as determined by SDS-PAGE. |
84869 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |