missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Caprin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Caprin-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7542-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049280

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
16-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_005889.3
Caprin-1 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
4076
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CAPRIN1
Recombinant Protein
E. coli
PSATSHSGSG SKSSGPPPPS GSSGSEAAAG AGAAAPASQH PATGTGAVQT EAMKQILGVI DKKLRNLEKK KGKLDDYQER MNKGERLNQD QLDAVSKYQE VTNNLEFAKE LQRSFMALSQ DIQKTIKKTA RREQLMREEA EQKRLKTVLE LQYVLDKLGD DEVRTDLKQG LNGVPILSEE ELSLLDEFYK LVDPERDMSL RLNEQYEHAS IHLWDLLEGK EKPVCGTTYK VLKEIVERVF QSNYFDSTHN HQNGLCEEEE AASAPAVEDQ VPEAEPEPAE EYTEQSEVES TEYVNRQFMA ETQFTSGEKE QVDEWTVETV EVVNSLQQQP QAASPSVPEP HSLTPVAQAD PLVRRQRVQD LMAQMQGPYN FIQDSMLDFE NQTLDPAIVS AQPMNPTQNM DMPQLVCPPV HSESRLAQPN QVPVQPEATQ VPLVSSTSEG YTASQPLYQP SHATEQRPQK EPIDQIQATI SLNTDQTTAS SSLPAASQPQ VFQAGTSKPL HSSGINVNAA PFQSMQTVFN MNAPVPPVNE PETLKQQNQY QASYNQSFSS QPHQVEQTEL QQEQLQTVVG TYHGSPDQSH QVTGNHQQPP QQNTGFPRSN QPYYNSRGVS RGGSRGARGL MNGYRGPANG FRGGYDGYRP SFSNTPNSGY TQSQFSAPRD YSGYQRDGYQ QNFKRGSGQS GPRGAPRGRG GPPRPNRGMP QMNTQQVN
Videos
Documentos