missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAPG / AFCP Protein
A DNA sequence encoding the Homo sapiens (Human) CAPG / AFCP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5753-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001243068.1 | |
CAPG / AFCP Protein | |
Research Use Only | |
CAPG | |
Recombinant Protein | |
E. coli | |
MYTAIPQSGS PFPGSVQDPG LHVWRVEKLK PVPVAQENQG VFFSGDSYLV LHNGPEEVSH LHLWIGQQSS RDEQGACAVL AVHLNTLLGE RPVQHREVQG NESDLFMSYF PRGLKYQEGG VESAFHKTST GAPAAIKKLY QVKGKKNIRA TERALNWDSF NTGDCFILDL GQNIFAWCGG KSNILERNKA RDLALAIRDS ERQGKAQVEI VTDGEEPAEM IQVLGPKPAL KEGNPEEDLT ADKANAQAAA LYKVSDATGQ MNLTKVADSS PFALELLISD DCFVLDNGLC GKIYIWKGRK ANEKERQAAL QVAEGFISRM QYAPNTQVEI LPQGRESPIF KQFFKDWK | |
Greater than 80% as determined by SDS-PAGE. |
822 | |
500 μg | |
Not determined. | |
Full Length of BC014549 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |