missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cancer / testis antigen 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Cancer/testis antigen 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5883-ye-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918439

  • 530.00€ / 100 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_066274.2
Cancer / testis antigen 2 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
30848
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CTAG2
Recombinant Protein
Yeast
MQAEGQGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPRGGA PRGPHGGAAS AQDGRCPCGA RRPDSRLLQL HITMPFSSPM EAELVRRILS RDAAPLPRPG AVLKDFTVSG NLLFMSVRDQ DREGAGRMRV VGWGLGSASP EGQKARDLRT PKHKVSEQRP GTPGPPPPEG AQGDGCRGVA FNVMFSAPHI
Videos