missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAMP Protein
A cDNA sequence encoding the CAMP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11270-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
820 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQVLSYKE AVLRAIDGIN QRSSDANLYR LLDLDPRPTM DGDPDTPKPV SFTVKETVCP RTTQQSPEDC DFKKDGLVKR CMGTVTLNQA RGSFDISCDK DNKRFALLGD FFRKSKEKIG KEFKRIVQRI KDFLRNLVPR TES | |
Greater than 90.0% as determined by SDS-PAGE. |
CAMP Protein | |
Research Use Only | |
CAMP | |
Human | |
His | |
CAMP protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |