missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Calpastatin Protein

A DNA sequence encoding the Homo sapiens (Human) Calpastatin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5765-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917609

  • 1453.00€ / 500 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

NP_001035905.1
Calpastatin Protein
500 μg
Not determined.
Full Length of isoform 4
Human
His
Tris-based buffer, 50% glycerol
831
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CAST
Recombinant Protein
Yeast
MNPTETKAVK TEPEKKSQST KPKSLPKQAS DTGSNDAHNK KAVSRSAEQQ PSEKSTEPKT KPQDMISAGG ESVAGITAIS GKPGDKKKEK KSLTPAVPVE SKPDKPSGKS GMDAALDDLI DTLGGPEETE EENTTYTGPE VSDPMSSTYI EELGKREVTI PPKYRELLAK KEGITGPPAD SSKPIGPDDA IDALSSDFTC GSPTAAGKKT EKEESTEVLK AQSAGTVRSA APPQEKKRKV EKDTMSDQAL EALSASLGTR QAEPELDLRS IKEVDEAKAK EEKLEKCGED DETIPSEYRL KPATDKDGKP LLPEPEEKPK PRSESELIDE LSEDFDRSEC KEKPSKPTEK TEESKAAAPA PVSEAVCRTS MCSIQSAPPE PATLKGTVPD DAVEALADSL GKKEADPEDG KPVMDKVKEK AKEEDREKLG EKEETIPPDY RLEEVKDKDG KPLLPKESKE QLPPMSEDFL LDALSEDFSG PQNASSLKFE DAKLAAAISE VVSQTPASTT QAGAPPRDTS QSDKDLDDAL DKLSDSLGQR QPDPDENKPM EDKVKEKAKA EHRDKLGERD DTIPPEYRHL LDDNGQDKPV KPPTKKSEDS KKPADDQDPI DALSGDLDSC PSTTETSQNT AKDKCKKAAS SSKAPKNGGK AKDSAKTTEE TSKPKDD
Videos