missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CACYBP Protein

A cDNA sequence encoding the CACYBP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11253-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916628

  • 103.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 03-04-2019
Añadir a la cesta

Especificación

Especificación

CACYBP Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CACYBP Human solution containing 20mM Trsi HCL pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQQKSQKKAE LLDNEKPAAV VAPITTGYTV KISNYGWDQS DKFVKIYITL TGVHQVPTEN VQVHFTERSF DLLVKNLNGK SYSMIVNNLL KPISVEGSSK KVKTDTVLIL CRKKVENTRW DYLTQVEKEC KEKEKPSYDT ETDPSEGLMN VLKKIYEDGD DDMKRTINKA WVESREKQAK GDTEF
Videos