missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CA13 Protein

A cDNA sequence encoding the CA13 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10542-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914198

  • 5941.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Especificación

Especificación

377677
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
CA13
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSP MGSMSRLSWG YREHNGPIHW KEFFPIADGD QQSPIEIKTK EVKYDSSLRP LSIKYDPSSA KIISNSGHSF NVDFDDTENK SVLRGGPLTG SYRLRQVHLH WGSADDHGSE HIVDGVSYAA ELHVVHWNSD KYPSFVEAAH EPDGLAVLGV FLQIGEPNSQ LQKITDTLDS IKEKGKQTRF TNFDLLSLLP PSWDYWTYPG SLTVPPLLES VTWIVLKQPI NISSQQLAKF RSLLCTAEGE AAAFLVSNHR PPQPLKGRKV RASFH
CA13 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 2,500 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of 4-nitrophenyl acetate to 4-nitrophenol per minute at pH 7.5 at 37C.
Human
His
CA13 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4),10% glycerol and 1mM DTT.
Videos