missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C4c Protein

A cDNA sequence encoding the C4c was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11240-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916588

  • 518.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

C4c Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 99.0%.
Research Use Only
Recombinant Protein
Native Protein
The Human Complement C4c solution is in Sodium Phosphate buffer pH 7.0 containing 0.15M NaCl and 0.09% NaN3.
Videos