missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF6 Protein
A cDNA sequence encoding the C1QTNF6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11233-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
C1QTNF6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS TFDRAVASGC QRCCDSEDPL DPAHVSSASS SGRPHALPEI RPYINITILK GDKGDPGPMG LPGYMGREGP QGEPGPQGSK GDKGEMGSPG APCQKRFFAF SVGRKTALHS GEDFQTLLFE RVFVNLDGCF DMATGQFAAP LRGIYFFSLN VHSWNYKETY VHIMHNQKEA VILYAQPSER SIMQSQSVML DLAYGDRVWV RLFKRQRENA IYSNDFDTYI TFSGHLIKAEDD | |
Greater than 95% as determined by SDS PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
0.23 mg/ml in 30mM Acetate Buffer pH 4. |