missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C14orf3, homolog 1 Protein

A DNA sequence encoding the Homo sapiens (Human) C14orf3, homolog 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8507-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055011

  • 675.00€ / 200 microgramos


Fecha estimada de envío
20-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001308370.1
C14orf3, homolog 1 Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
10598
Greater than 90% as determined by SDS-PAGE.
Research Use Only
C14orf3
Recombinant Protein
E. coli
MAKWGEGDPR WIVEERADAT NVNNWHWTER DASNWSTDKL KTLFLAVQVQ NEEGKCEVTE VSKLDGEASI NNRKGKLIFF YEWSVKLNWT GTSKSGVQYK GHVEIPNLSD ENSVDEVEIS VSLAKDEPDT NLVALMKEEG VKLLREAMGI YISTLKTEFT QGMILPTMNG ESVDPVGQPA LKTEERKAKP APSKTQARPV GVKIPTCKIT LKETFLTSPE ELYRVFTTQE LVQAFTHAPA TLEADRGGKF HMVDGNVSGE FTDLVPEKHI VMKWRFKSWP EGHFATITLT FIDKNGETEL CMEGRGIPAP EEERTRQGWQ RYYFEGIKQT FGYGARLF
Videos
Documentos