missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-X-C chemokine receptor type 4 Protein

A DNA sequence encoding the Homo sapiens (Human) C-X-C chemokine receptor type 4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9950-iv-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049071

  • 1865.00€ / 100 microgramos


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001008540.1
C-X-C chemokine receptor type 4 Protein
100 μg
Not determined.
Full Length Transmembrane Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
7852
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CXCR4
Recombinant Protein
Invitro-E. coli
MEGISIYTSD NYTEEMGSGD YDSMKEPCFR EENANFNKIF LPTIYSIIFL TGIVGNGLVI LVMGYQKKLR SMTDKYRLHL SVADLLFVIT LPFWAVDAVA NWYFGNFLCK AVHVIYTVNL YSSVLILAFI SLDRYLAIVH ATNSQRPRKL LAEKVVYVGV WIPALLLTIP DFIFANVSEA DDRYICDRFY PNDLWVVVFQ FQHIMVGLIL PGIVILSCYC IIISKLSHSK GHQKRKALKT TVILILAFFA CWLPYYIGIS IDSFILLEII KQGCEFENTV HKWISITEAL AFFHCCLNPI LYAFLGAKFK TSAQHALTSV SRGSSLKILS KGKRGGHSSV STESESSSFH SS
Videos
Documentos